missing translation for 'onlineSavingsMsg'
Learn More

Pumaprazole, MedChemExpress

Product Code. 16487708
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487708 10 mg 10mg
16411789 1 mg 1mg
16400178 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487708 Supplier Medchem Express Supplier No. HY19223
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pumaprazole is a reversible proton pump antagonist.

Pumaprazole is a reversible proton pump antagonist.

Specifications

Chemical Name or Material Pumaprazole
CAS 158364-59-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H22N4O2
Quantity 10 mg
Synonym BY-841
Solubility Information DMSO : 100 mg/mL (295.51 mM; Need ultrasonic)
SMILES O=C(OC)NC1=CC=CC(C)=C1CNC2=CC=CN3C2=NC(C)=C3C
Molecular Weight (g/mol) 338.4
Formula Weight 338.4
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.