missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUMA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£161.00 - £367.00
Specifications
| Antigen | PUMA |
|---|---|
| Concentration | 5.65 mg/mL |
| Dilution | Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274770
|
Novus Biologicals
NB500-261SS |
0.025 mL |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204870
|
Novus Biologicals
NB500-261 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PUMA Polyclonal specifically detects PUMA in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PUMA | |
| Western Blot 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Signal Transduction, Tumor Suppressors | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994 | |
| BBC3 | |
| IgG | |
| Affinity Purified | |
| 16 kDa |
| 5.65 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9BXH1 | |
| 27113 | |
| A synthetic peptide made to a region within the C-terminus of the human PUMA protein sequence (between residues 150-193). [UniProt# Q9BXH1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title