missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUMA alpha Monoclonal Antibody (2A8F6)

Product Code. 12827782
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12827782 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12827782 Supplier Invitrogen™ Supplier No. MA515752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

A suggested positive control is K562 cell lysate.

PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. Antisense inhibition of PUMA expression reduced the apoptotic response to p53, and PUMA is likely to play a role in mediating p53-induced cell death through the cytochrome c/Apaf-1-dependent pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PUMA alpha
Applications Western Blot
Classification Monoclonal
Clone 2A8F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene BBC3
Gene Accession No. Q80ZG6, Q9BXH1
Gene Alias BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1
Gene Symbols BBC3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 76 - 170 of human PUMA-alpha.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27113, 317673
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.