missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17043
This item is not returnable.
View return policy
Description
PUM3 Polyclonal specifically detects PUM3 in Human samples. It is validated for Western Blot.
Specifications
| KIAA0020 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| HBV X-transactivated gene 5 protein, HLA-HA8MGC8749, KIAA0020, KIAA0020 protein, Minor histocompatibility antigen HA-8, PEN, penguin homolog, protein 5 transactivated by hepatitis B virus X antigen (HBxAg), PUF6, XTP5HBV XAg-transactivated protein 5 | |
| Rabbit | |
| 74 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000 | |
| Q15397 | |
| KIAA0020 | |
| Recombinant protein encompassing a sequence within the center region of human KIAA0020. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9933 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction