missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM2, Mouse anti-Human, Clone: 5B6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00023369-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: DIMPSGRSRLLEDFRNNRFPNLQLRDLIGHIVEFSQDQHGSRFIQQKLERATPAERQMVFNEILQAAYQLMTDVFGNYVIQKFFEFGSLDQKLALATRSpecifications
| PUM2 | |
| Monoclonal | |
| Unconjugated | |
| pumilio homolog 2 (Drosophila) | |
| FLJ36528/KIAA0235/MGC138251/MGC138253/PUMH2/PUML2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 5B6 | |
| In 1x PBS, pH 7.4 | |
| NM_015317 | |
| PUM2 | |
| PUM2 (NP_056132, 701 a.a. ∼ 798 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 23369 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction