missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUM2 Monoclonal Antibody (740610)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561812

Product Code. 30536138

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PUM2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q8TB72
PUM2
E. coli-derived recombinant human PUM2, Pro103-Gly250.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
740610
Unconjugated
PUM2
5730503J23Rik; FLJ36528; KIAA0235; MGC138251; MGC138253; mKIAA0235; OTTHUMP00000200539; PUM1; Pum2; PUMH2; pumilio 2; pumilio homolog 2; pumilio RNA binding family member 2; pumilio RNA-binding family member 2; pumilio-2; PUML2; Pumm2
Mouse
Protein A/G
RUO
23369
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.