missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUM2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-387JF549
This item is not returnable.
View return policy
Description
PUM2 Polyclonal antibody specifically detects PUM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PUM2 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ36528, KIAA0235PUMH2PUML2, MGC138251, MGC138253, pumilio (Drosphila) homolog 2, pumilio homolog 2, pumilio homolog 2 (Drosophila), Pumilio-2 | |
| The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Pumilio 2 using the numbering given in entry NP_056132.1 (GeneID 23369). | |
| 0.1 mL | |
| Stem Cell Markers, Translation Control | |
| 23369 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction