missing translation for 'onlineSavingsMsg'
Learn More

Sphingosine Kinase 1/SPHK1 Antibody, R&D Systems™

Product Code. 18605954 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18605954 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605954 Supplier Novus Biologicals Supplier No. NBP323415
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Sphingosine Kinase 1/SPHK1 Polyclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1
Host Species Rabbit
Immunogen Sphingosine Kinase 1/SPHK1 synthetic peptide (coupled to carrier protein) corresponding to an unphosphorylated sequence of amino acids surrounding serine 225 in the central region of human Sphingosine Kinase 1/SPHK1. This sequence has 4 amino acid differences from mouse and 5 from rat Sphingosine Kinase 1/SPHK1, and is not homologous to sequences in Sphingosine Kinase 2/SPHK2. [UniProt# Q9NYA1]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8877
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.