Learn More
Erlab Pull-Down Shelf For Benchcap - Captair 481
Click to view available options
For Use With (Equipment):
Fuming Chamber
Unit Size:
Each
Description
- Pull-Down Shelf for Benchcap for your ductless fume hood Captair 481
- Intended Use: To store daily-use equipment
Specifications
Specifications
| For Use With (Equipment) | Fuming Chamber |
| Product Type | Shelf |