Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations
Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 0.9310g/mL |
| Boiling Point | 97°C (12.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 90% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4850 to 1.488 |
| Quantity | 500 g |
| Beilstein | 07, 81 |
| Merck Index | 15, 8046 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.