missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PUF60 Monoclonal Antibody (GT917)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531497

Product Code. 16374934

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PUF60 encodes a protein that is a Ro RNP-binding protein. It interacts with Ro RNPs and their interaction is thought to represent a gain of function for Ro RNPs. This protein also forms a ternary complex with far upstream element (FUSE) and FUSE-binding protein. It can repress a c-myc reporter via the FUSE. It is also known to target transcription factor IIH and inhibit activated transcription. This gene is implicated in the xeroderma pigmentosum disorder. There are two alternatively spliced transcript variants of this gene encoding different isoforms. There seems to be evidence of multiple polyadenylation sites for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PUF60
Monoclonal
1 mg/mL
PBS with no preservative
Q3UEB3, Q9UHX1, Q9WV25
Puf60
Recombinant protein encompassing a sequence within the center region of human PUF60. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
GT917
Unconjugated
Puf60
2410104I19Rik; 2810454F19Rik; 60 kDa poly(U)-binding-splicing factor; FBP interacting repressor; FBP-interacting repressor; FIR; FLJ31379; FUSE-binding protein-interacting repressor; LOW QUALITY PROTEIN: poly(U)-binding-splicing factor PUF60; poly(U) binding splicing factor 60; poly(U) binding splicing factor 60KDa; poly(U)-binding-splicing factor PUF60; poly-U binding splicing factor 60; poly-U binding splicing factor 60K; poly-U binding splicing factor 60KDa; PUF60; pyrimidine tract binding splicing factor; RNA-binding protein Siah-BP; Ro ribonucleoprotein-binding protein 1; Ro-binding protein 1; RoBP1; ROBPI; Siah binding protein 1; siah binding protein 1; FBP interacting repressor; pyrimidine tract binding splicing factor; Ro ribonucleoprotein-binding protein 1; siah-binding protein 1; SIAHBP1; Siah-BP1; similar to fuse-binding protein-interacting repressor isoform a; VRJS
Mouse
Protein G
RUO
22827, 67959, 84401
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.