missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUF60 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1125
This item is not returnable.
View return policy
Description
PUF60 Polyclonal specifically detects PUF60 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| PUF60 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FBP interacting repressor, FBP-interacting repressor, FIRpoly-U binding splicing factor PUF60, FLJ31379, FUSE-binding protein-interacting repressor, poly(U)-binding-splicing factor PUF60, poly-U binding splicing factor 60KDa, Ro ribonucleoprotein binding protein 1, Ro ribonucleoprotein-binding protein 1, Ro-binding protein 1, roBP1, ROBPI, siah binding protein 1,60 kDa poly(U)-binding-splicing factor, Siah-binding protein 1, siah-BP1, SIAHBP1pyrimidine tract binding splicing factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 22827 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| NP_055096 | |
| PUF60 | |
| Peptide with sequence C-YDQERFDNSDLSA corresponding to C-Terminus according to NP_510965.1, NP_055096.2, NP_001129505.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize all reported human isoforms (as represented by NP_510965.1; NP_055096.2; NP_001129505.1) | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction