missing translation for 'onlineSavingsMsg'
Learn More

Puerarin, MedChemExpress

Product Code. 16474767
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474767 50 mg 50mg
16448878 10 mg 10mg
16419006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16474767 Supplier Medchem Express Supplier No. HYN0145
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Puerarin, an isoflavone extracted from Radix puerariae, is a 5-HT2C receptor antagonist.

Puerarin, an isoflavone extracted from Radix puerariae, is a 5-HT2C receptor antagonist.

Specifications

Chemical Name or Material Puerarin
CAS 3681-99-0
Color White
Purity Grade Notes Research
Molecular Formula C21H20O9
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (120.08 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(O)C=C2)=COC3=C([C@H]4[C@@H]([C@H]([C@@H]([C@@H](CO)O4)O)O)O)C(O)=CC=C13
Molecular Weight (g/mol) 416.38
Formula Weight 416.38
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.