missing translation for 'onlineSavingsMsg'
Learn More

Puerarin, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66528.06

Quantity 5g

 View more versions of this product

Product Code. 15437929

  • £130.00 / 5g
 Restricted Item
Estimated Shipment: 04-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

3681-99-0
416.38
HKEAFJYKMMKDOR-VPRICQMDSA-N
53384442
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1
C21H20O9
MFCD00063399
8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone
7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one

Specifications

3681-99-0
MFCD00063399
64198
Soluble in DMSO or DMF; Slightly soluble in water or ethanol
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C1=C2OC=C(C(=O)C2=CC=C1O)C1=CC=C(O)C=C1
416.38
416.38
Puerarin
C21H20O9
5g
8-beta-d-glucopyranosyl-4',7-dihydroxyisoflavone
HKEAFJYKMMKDOR-VPRICQMDSA-N
7-hydroxy-3-(4-hydroxyphenyl)-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-4H-chromen-4-one
53384442
98%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.