missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PU.1 (SPI-1) (B-Cell Marker) Mouse, Clone: PU1/2146, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £403.13
Specifications
| Antigen | PU.1 (SPI-1) (B-Cell Marker) |
|---|---|
| Clone | PU1/2146 |
| Concentration | 200μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15930898
|
EnquireBio
6688-MSM2-P0 |
20 μg | |||||||
|
15940898
|
EnquireBio
6688-MSM2-P1 |
100 μg |
£425.41 £403.13 / 100µg Save £22.28 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| PU.1 (SPI-1) (B-Cell Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment (apprx. aa 16-170) of human PU.1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 40kDa |
| PU1/2146 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 6688 | |
| IgG2b κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear | |
| K-562 Cells, THP-1 cells. Lymph Node, Spleen, Hodgkin's Lymphoma, Colon Carcinoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title