missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PU.1/Spi-1 Antibody (PU1/2146), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £392.00
Specifications
| Antigen | PU.1/Spi-1 |
|---|---|
| Clone | PU1/2146 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2 μg/mL, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655358
|
Novus Biologicals
NBP2-75738-20ug |
20 μg |
£232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656267
|
Novus Biologicals
NBP2-75738-100ug |
100 μg |
£392.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PU.1/Spi-1 Monoclonal antibody specifically detects PU.1/Spi-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| PU.1/Spi-1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
| Recombinant fragment (around aa 16-170) of human PU.1/Spi-1 protein (Exact sequence is proprietary) (Uniprot: P17947 ) | |
| Primary | |
| Store at 4°C. |
| PU1/2146 | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry 1-2 μg/mL, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P17947 | |
| 6688 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title