missing translation for 'onlineSavingsMsg'
Learn More

PU.1/Spi-1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NBP1-00135

Product Code. 18252780

  • £398.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PU.1/Spi-1 Polyclonal specifically detects PU.1/Spi-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PU.1/Spi-1
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
hematopoietic transcription factor PU.1, OF, PU.1, SFPI131 kDa transforming protein, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1
Goat
Affinity Purified
RUO
Primary
This antibody is expected to recognize both reported isoforms (NP_001074016.1; NP_003111.2).
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 0.03 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:32000
NP_001074016.1
SPI1
Peptide with sequence C-DLYQRQTHEYY corresponding to internal region according to NP_001074016.1, NP_003111.2.
0.1 mg
Innate Immunity
6688
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only