missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564410
This item is not returnable.
View return policy
Description
Immunogen sequence: GFDETLAFSG RLTGFNIWDS VLSNEEIRET GGAESCHIRG NIVGWGVTEI QPHGGAQYVS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 90%.
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of long pentraxins, which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
Specifications
| PTX3 | |
| Polyclonal | |
| Unconjugated | |
| PTX3 | |
| AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P26022 | |
| PTX3 | |
| Recombinant protein corresponding to Human PTX3. Recombinant protein control fragment (Product #RP-104184). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction