Learn More
Invitrogen™ PTX3 Monoclonal Antibody (2C3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533376
Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: Western blot- A non-reduced sample treatment and SDS-Page was used. The band size is 400 kDa. Flow cytometry - This antibody stains the intra and extracellular domain of Pentraxin 3. For intracellular staining cells were permeabilized with buffer containing triton. Transfected cells were used as a positive control.
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of long pentraxins, which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
Specifications
| PTX3 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and no preservative | |
| P48759 | |
| PTX3 | |
| Recombinant mouse PTX3. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2C3 | |
| Unconjugated | |
| PTX3 | |
| AI607804; long pentraxin 3; pentaxin related; pentaxin-related protein PTX3; pentraxin 3; pentraxin 3, long; pentraxin related; pentraxin related gene; pentraxin-related protein PTX3; Ptx3; TNF alpha-induced protein 5; TNFAIP5; Tsg14; TSG-14; Tumor necrosis factor alpha-induced protein 5; tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14 | |
| Mouse | |
| Protein G | |
| RUO | |
| 19288 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.