missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTTG1IP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12575-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a single-pass type I integral membrane protein, which binds to pituitary tumor-transforming 1 protein (PTTG1), and facilitates translocation of PTTG1 into the nucleus. Coexpression of this protein and PTTG1 induces transcriptional activation of basic fibroblast growth factor. Alternatively spliced transcript variants have been found for this gene.Specifications
| PTTG1IP | |
| Polyclonal | |
| Unconjugated | |
| PTTG1IP | |
| C21orf1, C21orf3, PBF, PTTG binding factor, PTTG1IP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 754 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53801 | |
| PTTG1IP | |
| PTTG1IP Fusion Protein Ag3250 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion