missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTTG1IP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12575-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a single-pass type I integral membrane protein, which binds to pituitary tumor-transforming 1 protein (PTTG1), and facilitates translocation of PTTG1 into the nucleus. Coexpression of this protein and PTTG1 induces transcriptional activation of basic fibroblast growth factor. Alternatively spliced transcript variants have been found for this gene.Specifications
| PTTG1IP | |
| Polyclonal | |
| Unconjugated | |
| PTTG1IP | |
| C21orf1, C21orf3, PBF, PTTG binding factor, PTTG1IP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 754 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53801 | |
| PTTG1IP | |
| PTTG1IP Fusion Protein Ag3250 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction