missing translation for 'onlineSavingsMsg'
Learn More

PTTG1 Antibody (3F9), Novus Biologicals™

Product Code. 18378808 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378808 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378808 Supplier Novus Biologicals Supplier No. H00009232M11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTTG1 Monoclonal antibody specifically detects PTTG1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTTG1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3F9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004210
Gene Alias EAP1MGC126883, Esp1-associated protein, ESP1-associated protein 1, hPTTG, pituitary tumor-transforming 1, Pituitary tumor-transforming gene 1 protein, PTTGMGC138276, securin, Tumor-transforming protein 1, TUTR1HPTTG
Host Species Mouse
Immunogen PTTG1 (NP_004210, 93 a.a. ~ 113 a.a) synthetic peptide. MTEKTVKAKSSVPASDDAYPE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9232
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.