missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PTS Polyclonal specifically detects PTS in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | PTS |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | 6-pyruvoyl tetrahydrobiopterin synthase, 6-pyruvoyltetrahydropterin synthase, EC 4.2.3.12, FLJ97081, PTPSPTP synthase |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant PTS (Q03393; Met1-Glu145). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?