missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTRH1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94336-0.02ml
This item is not returnable.
View return policy
Description
PTRH1 Polyclonal antibody specifically detects PTRH1 in Human samples. It is validated for Western Blot
Specifications
| PTRH1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C9orf115, chromosome 9 open reading frame 115, EC 3.1.1.29, MGC51999, peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae), probable peptidyl-tRNA hydrolase, PTH, PTH1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-214 of human PTRH1 (NP_001002913.1). MRPGGFLGAGQRLSRAMSRCVLEPRPPGKRWMVAGLGNPGLPGTRHSVGMAVLGQLARRLGVAESWTRDRHCAADLALAPLGDAQLVLLRPRRLMNANGRSVARAAELFGLTAEEVYLVHDELDKPLGRLALKLGGSARGHNGVRSCISCLNSNAMPRLRVGIGRPAHPEAVQAHVLGCFSPAEQELLPLLLDRATDLILDHIRERSQGPSLGP | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 138428 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction