missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTRF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584948
This item is not returnable.
View return policy
Description
Immunogen sequence: VSVNVKTVRG SLERQAGQIK KLEVNEAELL RRRNFKVMIY QDEVKLPAKL SISKSLKESE ALPEKEGEEL GEGERPEEDA AALELSSD Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 94%.
The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3' end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
Specifications
| PTRF | |
| Polyclonal | |
| Unconjugated | |
| CAVIN1 | |
| 2310075E07Rik; AW546441; caveolae associated protein 1; caveolae-associated protein 1; CAVIN; CAVIN1; cavin-1; Cav-p60; CGL4; FKSG13; FLJ90031; Polymerase I and transcript release factor; PTRF; ptrf {ECO:0000312; RGD:1307685}; RNA polymerase I and transcript release factor; TTF-I interacting peptide 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NZI2 | |
| CAVIN1 | |
| Recombinant protein corresponding to Human CAVIN1. Recombinant protein control fragment (Product #RP-107984). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction