missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRT Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148743
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracellular catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP domain, Ig-like and fibronectin type III-like repeats. The protein domain structure and the expression pattern of the mouse counterpart of this PTP suggest its roles in both signal transduction and cellular adhesion in the central nervous system. Two alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported.
Specifications
| PTPRT | |
| Polyclonal | |
| Unconjugated | |
| PTPRT | |
| KIAA0283; mKIAA0283; mRPTPrho; protein tyrosine phosphatase, receptor type T; protein tyrosine phosphatase, receptor type, T; PTPRT; receptor protein tyrosine phosphatase; receptor protein tyrosine phosphatase-rho; receptor-type tyrosine-protein phosphatase rho; Receptor-type tyrosine-protein phosphatase T; RP5-1121H13.2; RPTPmam4; RPTP-rfo; RPTPrho; RPTP-rho; R-PTP-T | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O14522 | |
| PTPRT | |
| Mouse myeloma cell line NS0-derived recombinant human PTP rho /PTPRT, Gln26-Lys725. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction