missing translation for 'onlineSavingsMsg'
Learn More

PTPRO Antibody (3F4), Novus Biologicals™

Product Code. 18389578 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389578 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389578 Supplier Novus Biologicals Supplier No. H00005800M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTPRO Monoclonal antibody specifically detects PTPRO in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRO
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA 1:100 to 1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH35960
Gene Alias EC 3.1.3.48, GLEPP1PTPROt, Glomerular epithelial protein 1, phosphotyrosine phosphatase U2, protein tyrosine phosphatase PTP-U2, Protein tyrosine phosphatase U2, protein tyrosine phosphatase, receptor type, O, PTPase U2, PTP-oc, PTP-U2glomerular epithelial protein-1, PTPU2osteoclastic transmembrane protein-tyrosine phosphatase, receptor-type protein tyrosine phosphatase O, receptor-type tyrosine-protein phosphatase O, R-PTP-O
Host Species Mouse
Immunogen PTPRO (AAH35960, 1 a.a. ~ 597 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGHLPTGIHGARRLLPLLWLFVLFKNATAFHVTVQDDNNIVVSLEASDVISPASVYVVKITGESKNYFFEFEEFNSTLPPPVIFKASYHGLYYIITLVVVNGNVVTKPSRSITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYSGVSHEPKQHRTAPYPPQNISVRIVNLNKNNWEEQSGNFPEESFMRSQDTIGKEKLFHFTEETPEIPSGNISSGWPDFNSSDYETTSQPYWWDSASAAPESEDEFVSVLPMEYENNSTLSETEKSTSGSFSFFPVQMILTWLPPKPPTAFDGFHIHIEREENFTEYLMVDEEAHEFVAELKEPGKYKLSVTTFSSSGSCETRKSQSAKSLSFYISPSGEWIEELTEKPQHVSVHVLSSTTALMSWTSSQENYNSTIVSVVSLTCQKQKESQRLEKQYCTQVNSSKPIIENLVPGAQYQVVIYLRKGPLIGPPSDPVTFAIVPTGIKDLMLYPLGPTAVVLSWTRPYLGVFRKYVVEMFYFNPATMTSEWTTYYEIAATVSLTASVVIFP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5800
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.