missing translation for 'onlineSavingsMsg'
Learn More

PTPRN2 Antibody (4B9), Novus Biologicals™

Product Code. 18367348 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367348 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367348 Supplier Novus Biologicals Supplier No. H00005799M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTPRN2 Monoclonal antibody specifically detects PTPRN2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002838
Gene Alias EC 3.1.3.48, IA-2beta, IAR/receptor-like protein-tyrosine phosphatase, ICAARIARPTPRP, Islet cell autoantigen-related protein, KIAA0387, phogrin, protein tyrosine phosphatase receptor pi, protein tyrosine phosphatase, receptor type, N polypeptide 2, receptor-type tyrosine-protein phosphatase N2, R-PTP-N2, tyrosine phosphatase IA-2 beta
Host Species Mouse
Immunogen PTPRN2 (NP_002838, 108 a.a. ~ 212 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ELADLPKTYLRRPEASSPARPSKHSVGSERRYSREGGAALANALRRHLPFLEALSQAPASDVLARTHTAQDRPPAEGDDRFSESILTYVAHTSALTYPPGPRTQL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5799
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.