missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTPRM Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142321

Product Code. 17868772

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Additional Information: This antibody is expected to recognise both reported isoforms (NP_001098714.1 and NP_002836.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP mu (MAM) domain, an Ig-like domain and four fibronectin type III-like repeats. This PTP has been shown to mediate cell-cell aggregation through the interaction with another molecule of this PTP on an adjacent cell. This PTP can interact with scaffolding protein RACK1/GNB2L1, which may be necessary for the downstream signaling in response to cell-cell adhesion. Alternative splicing results in multiple transcripts encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTPRM
Polyclonal
Unconjugated
PTPRM
hR-PTPu; Kiaa4044; MGC166994; mKIAA4044; protein tyrosine phosphatase mu; protein tyrosine phosphatase receptor type M; protein tyrosine phosphatase receptor-type M; protein tyrosine phosphatase, receptor type M; protein tyrosine phosphatase, receptor type, M; protein tyrosine phosphatase, receptor type, mu polypeptide; protein tyrosine phosphatase, receptor-type, M; protein-tyrosine phosphatase mu; PTP mu; Ptpmu; PTPRL1; PTPRM; receptor-type tyrosine-protein phosphatase mu; RPTPM; RPTPmu; R-PTP-MU; RPTPU
Goat
Ammonium Sulfate Precipitation
RUO
5797
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P28827
PTPRM
Peptide with sequence C-KLIRQVDKWQEEYN.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.