missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563026
This item is not returnable.
View return policy
Description
Immunogen sequence: MTHMVNAMDR SYADQSTLHA EDPLSITFMD QHNFSPRYEN HSATAESSRL LDVPRYLCE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Protein tyrosine phosphatases, or PTPs, are type I transmembrane proteins, membrane associated proteins or proteins localized in nuclei. Examples of transmembrane PTPs are LAR, PTP alpha, PTP beta, PTP gamma, PTP delta, PTP epsilon, PTP zeta, PTPk and PTP-mu. Transmembrane PTPs play diverse roles during development and in adult tissues. Immunodepletion studies have suggested LAR to be a regulator of Insulin receptor phosphorylation. PTP alpha activity is increased twofold in response to phorbol ester stimulation, resulting in serine phosphorylation either directly or indirectly by members of the PKC family. Overexpression of v-H-ras and Neu, but not Myc or Int2, in mammary tumors has been shown to induce PTP epsilon expression. An alternative splicing event leads to a nervous tissue-specific chondroitin sulfate proteoglycan called phosphacan, which represents the amino terminal portion of PTP zeta. PTPk and PTP-mu share a conserved amino terminal 160 amino acid MAM domain which facilitates homophilic binding. PTP-mu localizes to points of cell contact and may be involved in regulating the assembly and disassembly of cadherin/catenin complexes in vivo.
Specifications
| PTPRK | |
| Polyclonal | |
| Unconjugated | |
| PTPRK | |
| AI853699; dJ480J14.2.1 (protein tyrosine phosphatase, receptor type, K (R-PTP-KAPPA, protein tyrosine phosphatase kappa , protein tyrosine phosphatase kappa; protein tyrosine phosphatase, receptor type K; protein tyrosine phosphatase, receptor type, K; protein tyrosine phosphatase, receptor type, K, extracellular region; protein-tyrosine phosphatase kappa; protein-tyrosine phosphatase, receptor type, kappa; Ptpk; PTPRK; receptor-like protein tyrosine phosphatase kappa extracellular region (RPTPK); receptor-type tyrosine-protein phosphatase kappa; Rptpk; R-PTPk; R-PTP-k; RPTPkappa; R-PTP-kappa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5796 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15262 | |
| PTPRK | |
| Recombinant protein corresponding to Human PTPRK. Recombinant protein control fragment (Product #RP-101384). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction