missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRH Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00229
This item is not returnable.
View return policy
Description
PTPRH Polyclonal specifically detects PTPRH in Human samples. It is validated for Peptide ELISA.
Specifications
| PTPRH | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| EC 3.1.3.48, FLJ39938, MGC133058, MGC133059, protein tyrosine phosphatase, receptor type, H, receptor-type tyrosine-protein phosphatase H, R-PTP-H, SAP1, SAP-1, Stomach cancer-associated protein tyrosine phosphatase 1, Transmembrane-type protein-tyrosine phosphatase type H | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5794 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:32000 | |
| NP_002833.2 | |
| PTPRH | |
| Peptide with sequence DHVRKNERDSNC corresponding to internal region according to NP_002833.2. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction