missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRF Monoclonal Antibody (6D7C7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538615
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Possible cell adhesion receptor. It possesses an intrinsic protein tyrosine phosphatase activity (PTPase).The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate.
Specifications
| PTPRF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P10586 | |
| PTPRF | |
| Purified recombinant fragment of human PTPRF (aa: extra 1104-1163) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 6D7C7 | |
| Unconjugated | |
| PTPRF | |
| AA591035; BNAH2; FLJ43335; FLJ45062; FLJ45567; LAR; LAR protein; LARFN5C; LARS; LCA homolog; LCA-homolog; leukocyte antigen-related (LAR) PTP receptor; leukocyte antigen-related tyrosine phosphatase; leukocyte common antigen related; protein tyrosine phosphatase receptor-type F; protein tyrosine phosphatase, receptor type F; protein tyrosine phosphatase, receptor type, F; protein tyrosine phosphatase, receptor type, F polypeptide; Ptprf; PTPRF protein; receptor-linked protein-tyrosine phosphatase LAR; receptor-type tyrosine-protein phosphatase F; RPTP-LAR | |
| Mouse | |
| Protein G | |
| RUO | |
| 5792 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction