missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPRCAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560487
This item is not returnable.
View return policy
Description
Immunogen sequence: DTDYDHVADG GLQADPGEGE QQCGEASSPE QVPVRAEEAR DSDTEGDLVL GSPGPASA Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 34%.
MSK-1 is a mitogen and stress activated protein kinase-1 which belongs to the AGC family of kinases and is related in structure to the ribosomal p70 S6 kinase subfamily. MSK-1 can be activated by ERK1/2 and SAPK2 /p38 MAP kinase. It is also known to be required for the phosphorylation of CREB, ATF1 H3 and HMG-14 in response to mitogen and stress. Similar to RSK, MSK-1 contains two kinase domains (N-term and a C-term). Once phosphorylated on Thr581 and Ser360 by ERK1/2 and SAPK2/p38, MSK-1 autophosphorylate on at least 5 sites. Of these autophosphorylation sites Ser212 and Ser376 get phosphorylated by the C-terminal kinase domain of MSK-1 which is essential for the catalytic activity of the N-terminal kinase domain.
Specifications
| PTPRCAP | |
| Polyclonal | |
| Unconjugated | |
| PTPRCAP | |
| AI605264; CD45 associated protein; CD-45-AP; CD45-AP; CD45-associated protein; LPAP; LSM-1; Lymphocyte phosphatase-associated phosphoprotein; protein tyrosine phosphatase receptor type C-associated protein; protein tyrosine phosphatase, receptor type C associated protein; protein tyrosine phosphatase, receptor type, C polypeptide-associated protein; protein tyrosine phosphatase, receptor type, C-associated protein; Ptprcap; PTPRC-associated protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14761 | |
| PTPRCAP | |
| Recombinant protein corresponding to Human PTPRCAP. Recombinant protein control fragment (Product #RP-100004). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction