missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRC Mouse anti-Human, Clone: HI30 (APC-C750), Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13943.100Reactions
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human PTPRC.
- High specificity
- High reactivity
Specifications
| PTPRC | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (5 uL/106 cells) Functional Study Immunocytochemistry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot The optimal w | |
| P08575 | |
| PTPRC | |
| Human PBMCs and tonsil cells. | |
| 100 Reactions | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| HI30 | |
| Mouse monoclonal antibody raised against human PTPRC. | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| B220/CD45/CD45R/GP180/LCA/LY5/T200 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 5788 | |
| Store in the dark at 4°C. Avoid prolonged exposure to light. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction