missing translation for 'onlineSavingsMsg'
Learn More

PTPRB Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-88829

Product Code. 18054934

  • £450.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PTPRB Polyclonal specifically detects PTPRB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PTPRB
Polyclonal
Unconjugated
DKFZp686E2262, DKFZp686H15164, EC 3.1.3.48, FLJ44133, HPTPB, HPTP-BETA, MGC142023, MGC59935, protein tyrosine phosphatase, receptor type, B, protein tyrosine phosphatase, receptor type, beta polypeptide, Protein-tyrosine phosphatase beta, PTPB, receptor-type tyrosine-protein phosphatase beta, R-PTP-BETA, Vascular endothelial protein tyrosine phosphatase, VEPTP, VE-PTP
Peptide with sequence C-ENHSFSQERTVPDK corresponding to internal region according to NP_001103224.1, NP_002828.3.
0.1 mg
Primary
Human
Purified
Flow Cytometry, Immunofluorescence, ELISA
0.5 mg/ml
Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:16000
Goat
Immunogen affinity purified
RUO
5787
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.