missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN7, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00005778-B02.50uL
This item is not returnable.
View return policy
Description
Sequence: MVQAHGGRSRAQPLTLSLGAAMTQPPPEKTPAKKHVRLQERRGSNVALMLDVRSLGAVEPICSVNTPREVTLHFLRTAGHPLTRWALQRQPPSPKQLEEEFLKIPSNFVSPEDLDIPGHASKDRYKTILPNPQSRVCLGRAQSQEDGDYINANYIRGYDGKEKVYIATQGPMPNTVSDFWEMVWQEEVSLIVMLTQLREGKEKCVHYWPTEEETYGPFQIRIQDMKECPEYTVRQLTIQYQEERRSVKHILFSAWPDHQTPESAGPLLRLVAEVEESPETAAHPGPIVVHCSAGIGRTGCFIATRIGCQQLKARGEVDILGIVCQLRLDRGGMIQTAEQYQFLHHTLALYAGQLPEEPSPSpecifications
| PTPN7 | |
| Polyclonal | |
| No additive | |
| NM_002832.2 | |
| PTPN7 | |
| PTPN7 (NP_002823.2, 1 a.a. ∼ 360 a.a) full-length human protein. | |
| RUO | |
| 5778 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| protein tyrosine phosphatase, non-receptor type 7 | |
| BPTP-4/HEPTP/LC-PTP/LPTP/PTPNI | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction