missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PTPN4 Antibody (2C8), Novus Biologicals™

Product Code. 18347548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347548 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347548 Supplier Novus Biologicals™ Supplier No. H00005775M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTPN4 Monoclonal antibody specifically detects PTPN4 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002821
Gene Alias EC 3.1.3.48, MEG, megakaryocyte phosphatase, megakaryocyte protein-tyrosine phosphatase, protein tyrosine phosphatase MEG1, protein tyrosine phosphatase, non-receptor type 4 (megakaryocyte), Protein-tyrosine phosphatase MEG1, PTPase-MEG1, PTPMEG, PTPMEG1, tyrosine-protein phosphatase non-receptor type 4
Host Species Mouse
Immunogen PTPN4 (NP_002821, 361 a.a. ∽ 460 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5775
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.