missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PTPN3 Polyclonal antibody specifically detects PTPN3 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PTPN3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000 - 1:25000 |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | DKFZp686N0569, EC 3.1.3.48, protein tyrosin phosphatase H1, protein tyrosine phosphatase, non-receptor type 3, Protein-tyrosine phosphatase H1, PTP-H1, PTPH1cytoskeletal-associated protein tyrosine phosphatase, tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 3 |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between between 175 and 225.. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?