Learn More
PTPN23 Rabbit, anti-Human, CoraLite 647, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL647-10472-20UL
 View more versions of this product
Product Code. 18006685
  • £135.00 / 20µL
Description
The PTPN23 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PTPN23. This antibody recognizes human antigen. The PTPN23 antibody has been validated for the following applications: IF analysis. This antibody is coralite™647.
Specifications
Specifications
PTPN23
1000 μg/mL
DKFZp564F0923, HD PTP, HDPTP, KIAA1471, PTP TD14, PTPN23
20 μL
25930
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite 647
Rabbit
Primary
Human
150 kDa