missing translation for 'onlineSavingsMsg'
Learn More

PTPN23 Antibody, Novus Biologicals™

Product Code. 18353829 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353829 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353829 Supplier Novus Biologicals Supplier No. H00025930B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PTPN23 Polyclonal antibody specifically detects PTPN23 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPN23
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH04881
Gene Alias EC 3.1.3.48, HDPTP, HD-PTPDKFZp564F0923, His domain-containing protein tyrosine phosphatase, His-domain protein tyrosine phosphatase, KIAA1471DKFZP564F0923, Protein tyrosine phosphatase TD14, protein tyrosine phosphatase, non-receptor type 23, PTP-TD14, tyrosine-protein phosphatase non-receptor type 23
Host Species Mouse
Immunogen PTPN23 (AAH04881, 1 a.a. - 577 a.a.) full-length human protein. MGPQAAPLTIRGPSSAGQSTPSPHLVPSPAPSPGPGPVPPRPPAAEPPPCLRRGAAAADLLSSSPESQHGGTQSPGGGQPLLQPTKVDAAEGRRPQALRLIERDPYEHPERLRQLQQELEAFRGQLGDVGALDTVWRELQDAQEHDARGRSIAIARCYSLKNRHQDVMPYDSNRVVLRSGKDDYINASCVEGLSPYCPPLVATQAPLPGTAADFWLMVHEQKVSVIVMLVSEAEMEKQKVARYFPTERGQPMVHGALSLALSSVRSTETHVERVLSLQFRDQSLKRSLVHLHFPTWPELGLPDSPSNLLRFIQEVHAHYLHQRPLHTPIIVHCSSGVGRTGAFALLYAAVQEVEAGNGIPELPQLVRRMRQQRKHMLQEKLHLRFCYEAVVRHVEQVLQRHGVPPPCKPLASASISQKNHLPQDSQDLVLGGDVPISSIQATIAKLSIRPPGGLESPVASLPGPAEPPGLPPASLPESTPIPSSSPPPLSSPLPEAPQPKEEPPVPEAPSSGPPSSSLELLASLTPEAFSLDSSLRGKQRMSKHNFLQAHNGQGLRATRPSDDPLSLLDPLWTLNKT
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25930
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.