missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN23 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36549R
This item is not returnable.
View return policy
Description
PTPN23 Polyclonal antibody specifically detects PTPN23 in Human samples. It is validated for Western Blot
Specifications
| PTPN23 | |
| Polyclonal | |
| Western Blot | |
| EC 3.1.3.48, HDPTP, HD-PTPDKFZp564F0923, His domain-containing protein tyrosine phosphatase, His-domain protein tyrosine phosphatase, KIAA1471DKFZP564F0923, Protein tyrosine phosphatase TD14, protein tyrosine phosphatase, non-receptor type 23, PTP-TD14, tyrosine-protein phosphatase non-receptor type 23 | |
| The immunogen recognized by the antibody maps to a region between residue 1586 to 1636 of human HisDomain Protein Tyrosine Phosphatase using the numbering given in entry NP_056281.1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 25930 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction