missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN21 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68974
This item is not returnable.
View return policy
Description
PTPN21 Polyclonal antibody specifically detects PTPN21 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PTPN21 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| dJ1175B15.2 (protein tyrosine phosphatase, non-receptor type 21), EC 3.1.3, EC 3.1.3.48, protein tyrosine phosphatase, non-receptor type 21, Protein-tyrosine phosphatase D1, PTPD1protein tyrosine phosphatase D1, PTPRL10, tyrosine-protein phosphatase non-receptor type 21 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KVALLHQKYRGLTAPDAEMLYMQEVERMDGYGEESYPAKDSQGSDISIGACLEGIFVKHKNGRHPVVFRWHDIANMSHNKS | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11099 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction