missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPN18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113316
This item is not returnable.
View return policy
Description
PTPN18 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
Specifications
| PTPN18 | |
| Polyclonal | |
| Unconjugated | |
| PTPN18 | |
| BDP1; Brain-derived phosphatase; fetal liver phosphatase 1; FLP1; FLP-1; HSCF; protein tyrosine phosphatase 20; protein tyrosine phosphatase, non-receptor type 18; protein tyrosine phosphatase, non-receptor type 18 (brain-derived); PTP20; PTP-HSCF; Ptpk1; PTP-K1; Ptpn18; Tyrosine-protein phosphatase non-receptor type 18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26469, 301333 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99952 | |
| PTPN18 | |
| Recombinant Human Tyrosine-protein phosphatase non-receptor type 18 protein (1-210AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction