Learn More
Invitrogen™ PTPN18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597080
Description
Immunogen sequence: MSRSLDSARS FLERLEARGG REGAVLAGEF SKRCERYWAQ EQEPLQTGLF CITLIKEKWL NEDIMLRTLK VTFQKESRSV YQLQYMSWPD RGVPSSPDHM LAMVEEARRL QGSGPEPLCV HCSAGCGRTG VLCTVDYVRQ LLLTQMIPPD FSLFDVVLKM RKQRPAAVQT EEQYRFLYHT VAQMFCSTLQ NASPHYQNIK ENCAPLYDDA; Positive Samples: U-251MG, SW620; Cellular Location: Cytoplasm, Nucleus.
Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
Specifications
| PTPN18 | |
| Polyclonal | |
| Unconjugated | |
| PTPN18 | |
| BDP1; Brain-derived phosphatase; fetal liver phosphatase 1; FLP1; FLP-1; HSCF; protein tyrosine phosphatase 20; protein tyrosine phosphatase, non-receptor type 18; protein tyrosine phosphatase, non-receptor type 18 (brain-derived); PTP20; PTP-HSCF; Ptpk1; PTP-K1; Ptpn18; Tyrosine-protein phosphatase non-receptor type 18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26469 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.98 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99952 | |
| PTPN18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human PTPN18 (NP_0011358421). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.