missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPN14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531285
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal noncatalytic domain similar to that of band 4.1 superfamily cytoskeleton-associated proteins, which suggested the membrane or cytoskeleton localization of this protein. The specific function of this PTP has not yet been determined.
Specifications
| PTPN14 | |
| Polyclonal | |
| Unconjugated | |
| PTPN14 | |
| C130080N23Rik; cytoskeletal-associated protein tyrosine phosphatase; OTTMUSG00000022087; PEZ; protein tyrosine phosphatase, non-receptor type 14; protein-tyrosine phosphatase pez; Protein-tyrosine phosphatase PTP36; PTP36; PTPD2; Ptpn14; Tyrosine-protein phosphatase non-receptor type 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5784 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15678 | |
| PTPN14 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Pez. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction