missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPN12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582281
This item is not returnable.
View return policy
Description
Immunogen sequence: SVTQSNKVSV TPPEESQNSD TPPRPDRLPL DEKGHVTWSF HGPENAIPIP DLSEGNSSDI NYQTRKTVSL TPSPTTQVET PDLVDHDNTS PLFRTPLSFT NPLHSDDSDS DERNSDGAVT QNKTNISTAS ATVSAAT Highest antigen sequence indentity to the following orthologs: Rat - 64%, Mouse - 63%.
PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
Specifications
| PTPN12 | |
| Polyclonal | |
| Unconjugated | |
| PTPN12 | |
| MPTP-PEST; P19-PTP; pest; protein tyrosine phosphatase non-receptor type 12; protein tyrosine phosphatase, non-receptor type 12; protein-tyrosine phosphatase G1; Protein-tyrosine phosphatase P19; PTPG1; PTPN12; PTP-P19; PTP-PEST; RKPTP; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5782 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q05209 | |
| PTPN12 | |
| Recombinant protein corresponding to Human PTPN12. Recombinant protein control fragment (Product #RP-89757). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction