missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN11, Rabbit anti-Human, Clone: 4A2, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19077.200uL
This item is not returnable.
View return policy
Specifications
| PTPN11 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA). | |
| BPTP3/CFC/MGC14433/NS1/PTP-1D/PTP2C/SH-PTP2/SH-PTP3/SHP2 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 4A2 | |
| Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user. | |
| protein tyrosine phosphatase, non-receptor type 11 | |
| PTPN11 | |
| A synthetic phosphopeptide corresponding to residues surrounding Y580 of human PTPN11. | |
| RUO | |
| 5781 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction