missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560557
This item is not returnable.
View return policy
Description
Immunogen sequence: IPTLDNLQKG VQFALKYQSL GQCVYVHCKA GRSRSATMVA AYLIQVHKWS PEEAVRAIAK IRSYIHIRPG QLDVLKEFHK QITARAT Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PGP is an essential intermediate in the biosynthetic pathway of cardiolipin, a mitochondrial-specific phospholipid regulating the membrane integrity and activities of the organelle. Has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. Probably involved in regulation of insulin secretion in pancreatic beta cells.
Specifications
| PTPMT1 | |
| Polyclonal | |
| Unconjugated | |
| PTPMT1 | |
| 1110001D10Rik; 2810004N20Rik; DUSP23; MOSP; NB4 apoptosis/differentiation related protein; phosphatidylglycerophosphatase and protein-tyrosine phosphatase 1; Phosphoinositide lipid phosphatase; Plip; PNAS-129; protein tyrosine phosphatase, mitochondrial 1; protein-tyrosine phosphatase mitochondrial 1; PTEN-like phosphatase; PTPMT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUK0 | |
| PTPMT1 | |
| Recombinant protein corresponding to Human PTPMT1. Recombinant protein control fragment (Product #RP-97310). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Dela en innehållskorrigering