missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPLB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524897
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
PTPLB is probable anti-phosphatase.
Specifications
| PTPLB | |
| Polyclonal | |
| Unconjugated | |
| Hacd2 | |
| 3-hydroxyacyl-CoA dehydratase 2; 6330408J20Rik; AI255777; AI481689; HACD2; Hcad2; I79_000662; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member b; protein-tyrosine phosphatase-like member B; Ptplb; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 201562 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6Y1H2 | |
| Hacd2 | |
| KLH conjugated synthetic peptide between 226-254 amino acids from the C-terminal region of human PTPLB. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction