missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTPLAD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566320
This item is not returnable.
View return policy
Description
Immunogen sequence: SIPIFNETGRF SFTLPYPVK Highest antigen sequence identity to the following orthologs - mouse 90%, rat 80%.
Catalyzes the third of the four reactions of the long-chain fatty acids elongation cycle. This endoplasmic reticulum-bound enzymatic process, allows the addition of two carbons to the chain of long- and very long-chain fatty acids/VLCFAs per cycle. This enzyme catalyzes the dehydration of the 3-hydroxyacyl-CoA intermediate into trans-2,3-enoyl-CoA, within each cycle of fatty acid elongation. Thereby, it participates in the production of VLCFAs of different chain lengths that are involved in multiple biological processes as precursors of membrane lipids and lipid mediators. May be involved in Rac1-signaling pathways leading to the modulation of gene expression. Promotes insulin receptor/INSR autophosphorylation and is involved in INSR internalization (PubMed:25687571).
Specifications
| PTPLAD1 | |
| Polyclonal | |
| Unconjugated | |
| HACD3 | |
| 3-hydroxyacyl-CoA dehydratase 3; BIND1; B-ind1; Butyrate-induced protein 1; butyrate-induced transcript 1; HACD3; HSPC121; protein tyrosine phosphatase-like A domain containing 1; protein tyrosine phosphatase-like protein PTPLAD1; protein-tyrosine phosphatase-like A domain-containing protein 1; PTPLAD1; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 3; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51495 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P035 | |
| HACD3 | |
| Recombinant Human PTPLAD1. Recombinant protein control fragment (Product #RP-107001). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction